3-Bromoimidazo[1,2-a]pyridine-6-carbaldehyde

Supplier: APOLLO SCIENTIFIC

Warning

OR110284-250MG OR110284-1G
76563-956EA 308 USD
76563-956 76563-954
3-Bromoimidazo[1,2-a]pyridine-6-carbaldehyde
3-Bromoimidazo[1,2-a]pyridine-6-carbaldehyde
Formula: C₈H₅BrN₂O
MW: 225.05
Melting Pt: 207…210 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD14583145
CAS Number: 1004550-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR