3-Bromo-6-chloroimidazo[1,2-a]pyridine

Supplier: APOLLO SCIENTIFIC

Warning

OR110278-1G OR110278-5G
76563-946EA 208 USD
76563-946 76563-944
3-Bromo-6-chloroimidazo[1,2-a]pyridine
3-Bromo-6-chloroimidazo[1,2-a]pyridine
Formula: C₇H₄BrClN₂
MW: 231.48 g/mol
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD08667858
CAS Number: 886371-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR