2-Amino-6-bromoquinazoline

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 6-Bromoquinazolin-2-amine

OR110259-1G
76563-918EA 76 USD
76563-918
2-Amino-6-bromoquinazoline
2-Amino-6-bromoquinazoline
Formula: C₈H₆BrN₃
MW: 224.0600 g/mol
Melting Pt: 270…272 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114089
CAS Number: 190273-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR