4-Pyridineboronic acid pinacol ester

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

OR11025-100G OR11025-25G
76561-060EA 207 USD
76561-060 76559-780
4-Pyridineboronic acid pinacol ester
4-Pyridineboronic acid pinacol ester
Formula: C₁₁H₁₆BNO₂
MW: 205.06 g/mol
Melting Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD01319051
CAS Number: 181219-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR