1H-Pyrazole-4-boronic acid pinacol ester

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

OR11023-5G OR11023-1G
76563-868EA 120 USD
76563-868 76563-866
1H-Pyrazole-4-boronic acid pinacol ester
1H-Pyrazole-4-boronic acid pinacol ester
Formula: C₉H₁₅BN₂O₂
MW: 194.039 g/mol
Melting Pt: 142…149 °C
Storage Temperature: Ambient
MDL Number: MFCD03453063
CAS Number: 269410-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR