2-Amino-7-bromoquinazoline

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 7-Bromoquinazolin-2-amine

OR110183-1G
76563-796EA 95 USD
76563-796
2-Amino-7-bromoquinazoline
2-Amino-7-bromoquinazoline
Formula: C₈H₆BrN₃
MW: 224.0600 g/mol
Melting Pt: 280…283 °C
Storage Temperature: Ambient
MDL Number: MFCD09027698
CAS Number: 190274-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR