Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid ethyl esterEthyl 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate

OR11018-5G OR11018-1G
76563-792EA 50 USD
76563-792 76563-794
Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate
Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate
Formula: C₁₄H₁₆N₂O₃
MW: 260.3 g/mol
Melting Pt: 208 °C
Storage Temperature: Ambient
MDL Number: MFCD00124778
CAS Number: 5395-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR