1-Methyl-6-nitro-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

OR110173-25G OR110173-1G OR110173-5G
76563-782EA 103 USD
76563-782 76563-780 76563-778
1-Methyl-6-nitro-1H-indazole
1-Methyl-6-nitro-1H-indazole
Formula: C₈H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00456124
CAS Number: 6850-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR