4-Hydroxyphenylboronic acid pinacol ester 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Hydroxybenzeneboronic acid pinacol ester4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

OR11014-5G OR11014-1G OR11014-25G
76563-714EA 49 USD
76563-714 76563-712 76563-710
4-Hydroxyphenylboronic acid pinacol ester 98%
4-Hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₇BO₃
MW: 220.08 g/mol
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD02093756
CAS Number: 269409-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR