3-Bromo-7-chloroimidazo[1,2-a]pyridine

Supplier: APOLLO SCIENTIFIC

Warning

OR110071-1G
76563-652EA 141 USD
76563-652
3-Bromo-7-chloroimidazo[1,2-a]pyridine
3-Bromo-7-chloroimidazo[1,2-a]pyridine
Formula: C₇H₄BrClN₂
MW: 231.4800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111834
CAS Number: 342613-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR