N-(4-Ethoxycarbonylphenyl)maleamic acid

Supplier: APOLLO SCIENTIFIC

Synonyms: 4-{[4-(Ethoxycarbonyl)phenyl]amino}-4-oxobut-2-enoic acid

OR110035-25G OR110035-5G
76563-622EA 86 USD
76563-622 77403-022
N-(4-Ethoxycarbonylphenyl)maleamic acid
N-(4-Ethoxycarbonylphenyl)maleamic acid
Formula: C₁₃H₁₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD00063173
CAS Number: 200126-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR