4-Iodo-1H-pyrazole 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Iodopyrazole

OR10990-25G OR10990-100G OR10990-500G
76563-584EA 13 USD
76563-584 76563-588 76563-586
4-Iodo-1H-pyrazole 99%
4-Iodo-1H-pyrazole
Formula: C₃H₃IN₂
MW: 193.97 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005244
CAS Number: 3469-69-0
EINECS: 222-434-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR