3-Bromo-4-hydroxybenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR1074-5G OR1074-100G OR1074-25G
76561-206EA 13 USD
76561-206 76560-602 76561-388
3-Bromo-4-hydroxybenzonitrile
3-Bromo-4-hydroxybenzonitrile
Formula: C₇H₄BrNO
MW: 198.02 g/mol
Melting Pt: 155…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00143096
CAS Number: 2315-86-8
EINECS: 219-022-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR