4-tert-Butoxyphenol 98%

Warning

Synonyms: Hydroquinone Mono-tert-butyl Ether

OR10627-5G
76561-942EA 145.23 USD
76561-942
4-tert-Butoxyphenol 98%
4-tert-Butoxyphenol
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD02183556
CAS Number: 2460-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR