1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1-(Phenylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

OR10602-1G OR10602-25G OR10602-5G
76561-522EA 68 USD
76561-522 76560-116 76562-072
1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester
1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₆H₂₁BN₂O₂
MW: 284.16 g/mol
Melting Pt: 86…91 °C
Storage Temperature: Ambient
EINECS: 261-911-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR