3-(Methylthio)phenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-(Methylthio)benzeneboronic acid3-Thioanisoleboronic acid

OR10567-25G OR10567-5G
76562-404EA 18 USD
76562-404 76560-254
3-(Methylthio)phenylboronic acid 97%
3-(Methylthio)phenylboronic acid
Formula: C₇H₉BO₂S
MW: 168.02 g/mol
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD01319105
CAS Number: 128312-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR