4-(Methylcarbamoyl)phenylboronic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-(N-Methylaminocarbonyl)phenylboronic acid4-(Methylcarbamoyl)benzeneboronic acid

OR10555-10G OR10555-1G OR10555-5G
76563-188EA 88 USD
76563-188 76561-158 76560-446
4-(Methylcarbamoyl)phenylboronic acid
4-(Methylcarbamoyl)phenylboronic acid
Formula: C₈H₁₀BNO₃
MW: 178.98 g/mol
Melting Pt: 200…220 °C
Storage Temperature: Ambient
MDL Number: MFCD03788427
CAS Number: 121177-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR