3-(Methylcarbamoyl)phenylboronic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-(Methylcarbamoyl)benzeneboronic acid

OR10554-1G OR10554-250MG
76560-606EA 22 USD
76560-606 76563-300
3-(Methylcarbamoyl)phenylboronic acid
3-(Methylcarbamoyl)phenylboronic acid
Formula: C₈H₁₀BNO₃
MW: 178.98
Melting Pt: 114…124 °C
Storage Temperature: Ambient
MDL Number: MFCD04038918
CAS Number: 832695-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR