Methyl tetrahydropyran-4-carboxylate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl tetrahydro-2H-pyran-4-carboxylate

OR10552-25G OR10552-500G OR10552-100G
76561-248EA 632 USD
76561-248 76560-376 76560-670
Methyl tetrahydropyran-4-carboxylate
Methyl tetrahydropyran-4-carboxylate
Formula: C₇H₁₂O₃
MW: 144.1684 g/mol
Boiling Pt: 196.8 °C
Melting Pt: –33 °C
Density: 1.08 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD01075170
CAS Number: 110238-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR