3-(Methoxycarbonyl)phenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-(Methoxycarbonyl)benzeneboronic acid

OR10536-1G OR10536-5G
76562-730EA 15 USD
76562-730 76563-164
3-(Methoxycarbonyl)phenylboronic acid 97%
3-(Methoxycarbonyl)phenylboronic acid
Formula: C₈H₉BO₄
MW: 179.97 g/mol
Melting Pt: 205…208 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093046
CAS Number: 99769-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR