4-Bromo-3-picoline hydrochloride

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Bromo-3-methylpyridine hydrochloride

OR1050-5G OR1050-250MG OR1050-1G
76560-984EA 20 USD
76560-984 76561-174 76560-026
4-Bromo-3-picoline hydrochloride
4-Bromo-3-picoline hydrochloride
Formula: C₆H₇BrClN
Storage Temperature: Ambient
MDL Number: MFCD04966778
CAS Number: 40899-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR