4-Hydroxy-2,6-dimethoxybenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,6-Dimethoxy-4-hydroxybenzaldehyde

OR10495-1G OR10495-25G OR10495-5G
76559-802EA 101 USD
76559-802 76560-358 76560-860
4-Hydroxy-2,6-dimethoxybenzaldehyde 98%
4-Hydroxy-2,6-dimethoxybenzaldehyde
Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
Melting Pt: 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01407645
CAS Number: 22080-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR