3-(Ethoxycarbonyl)phenylboronic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-(Ethoxycarbonyl)benzeneboronic acid3-Ethoxycarbonylphenylboronic acid

OR10469-25G OR10469-5G OR10469-100G
76563-272EA 15 USD
76563-272 76560-248 76561-990
3-(Ethoxycarbonyl)phenylboronic acid
3-(Ethoxycarbonyl)phenylboronic acid
Formula: C₉H₁₁BO₄
MW: 193.99 g/mol
Melting Pt: 131…139 °C
Storage Temperature: Ambient
MDL Number: MFCD02179444
CAS Number: 4334-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR