4-(Cyclohexylcarbamoyl)phenylboronic acid

Supplier: APOLLO SCIENTIFIC

Warning

OR10430-25G OR10430-5G OR10430-1G
76563-254EA 44 USD
76563-254 76562-788 76560-080
4-(Cyclohexylcarbamoyl)phenylboronic acid
4-(Cyclohexylcarbamoyl)phenylboronic acid
Formula: C₁₃H₁₈BNO₃
Melting Pt: 174…178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411956
CAS Number: 762262-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR