5-Chloro-2-methylphenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (5-Chloro-2-methylphenyl)boronic acid5-Chloro-2-methylbenzeneboronic acid

OR10415-5G OR10415-10G OR10415-1G
76559-694EA 37 USD
76559-694 76562-486 76563-156
5-Chloro-2-methylphenylboronic acid 97%
5-Chloro-2-methylphenylboronic acid
Formula: C₇H₈BClO₂
MW: 170.40 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD03411939
CAS Number: 148839-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR