4-Carboxy-2-nitrophenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Carboxy-2-nitrobenzeneboronic acid

OR10401-1G OR10401-250MG
76561-238EA 22.66 USD
76561-238 76561-724
4-Carboxy-2-nitrophenylboronic acid 97%
4-Carboxy-2-nitrophenylboronic acid
Formula: C₇H₆BNO₆
MW: 210.94 g/mol
Melting Pt: 265…268 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00448780
CAS Number: 85107-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR