4-Bromo-2-methylimidazole 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromo-2-methyl-1H-imidazole

OR10370-5G OR10370-25G
76560-428EA 76 USD
76560-428 76560-806
4-Bromo-2-methylimidazole 98%
4-Bromo-2-methylimidazole
Formula: C₄H₅BrN₂
MW: 161 g/mol
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD03411959
CAS Number: 16265-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR