4-(Benzyloxy)phenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: (4-(Benzyloxy)phenyl)boronic acid4-Benzyloxybenzeneboronic acid

OR10362-25G OR10362-5G OR10362-100G
76562-512EA 49 USD
76562-512 76560-270 76560-100
4-(Benzyloxy)phenylboronic acid 98%
4-(Benzyloxy)phenylboronic acid
Formula: C₁₃H₁₃BO₃
MW: 228.06 g/mol
Melting Pt: 198…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01075705
CAS Number: 146631-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR