4-(Benzylcarbamoyl)phenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Benzylcarbamoyl)benzeneboronic acid(4-(Benzylcarbamoyl)phenyl)boronic acid4-(N-Benzylaminocarbonyl)phenylboronic acid

OR10355-5G OR10355-1G
76560-180EA 23 USD
76560-180 76561-092
4-(Benzylcarbamoyl)phenylboronic acid 98%
4-(Benzylcarbamoyl)phenylboronic acid
Formula: C₁₄H₁₄BNO₃
MW: 255.08 g/mol
Melting Pt: 212…218 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411953
CAS Number: 252663-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR