4-Aminophenylboronic acid hydrochloride

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: (4-Aminophenyl)boronic acid hydrochloride4-Aminobenzeneboronic acid hydrochloride

OR10338-5G OR10338-1G OR10338-25G
76562-544EA 165 USD
76562-544 76561-800 76559-818
4-Aminophenylboronic acid hydrochloride
4-Aminophenylboronic acid hydrochloride
Formula: C₆H₉BClNO₂
MW: 173.41 g/mol
Melting Pt: 141…143 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03001333
CAS Number: 80460-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR