4-(Aminomethyl)phenylboronic acid hydrochloride 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Aminomethylphenylboronic acid hydrochloride4-(Aminomethyl)benzeneboronic acid hydrochloride

OR10330-1G OR10330-250MG
76562-896EA 19.48 USD
76562-896 76560-056
4-(Aminomethyl)phenylboronic acid hydrochloride 95%
4-(Aminomethyl)phenylboronic acid hydrochloride
Formula: C₇H₁₁BClNO₂
MW: 187.43 g/mol
Melting Pt: 240…250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01632199
CAS Number: 75705-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR