2,3,5-Tribromothiophene

Supplier: APOLLO SCIENTIFIC

Warning

OR10149-25G OR10149-100G OR10149-500G
76562-538EA 147 USD
76562-538 76563-280 76560-986
2,3,5-Tribromothiophene
2,3,5-Tribromothiophene
Formula: C₄HBr₃S
Boiling Pt: 120 °C/11mmHg
Melting Pt: 25…29 °C
Density: 2.483 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00014521
CAS Number: 3141-24-0
EINECS: 221-544-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR