Pyridazine-3-carboxylic acid 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR1005-5G OR1005-10G OR1005-1G
76563-264EA 47 USD
76563-264 76560-122 76559-848
Pyridazine-3-carboxylic acid 96%
Pyridazine-3-carboxylic acid
Formula: C₅H₄N₂O₂
MW: 124.10 g/mol
Melting Pt: 190…195 °C
Storage Temperature: Ambient
MDL Number: MFCD01646333
CAS Number: 2164-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR