3-Azetidinecarboxylic acid 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Azetidine-3-carboxylic acid3-Azetanecarboxylic acid

OR0961-25G OR0961-1G OR0961-5G
76562-334EA 16.4 USD
76562-334 76563-216 76562-750
3-Azetidinecarboxylic acid 96%
3-Azetidinecarboxylic acid
Formula: C₄H₇NO₂
MW: 101.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191763
CAS Number: 36476-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR