1-(3-Chlorophenyl)biguanide hydrochloride

Supplier: APOLLO SCIENTIFIC

Warning

OR0928-5G OR0928-25G
76561-024EA 141 USD
76561-024 76561-210
1-(3-Chlorophenyl)biguanide hydrochloride
1-(3-Chlorophenyl)biguanide hydrochloride
Formula: C₈H₁₁Cl₂N₅
Storage Temperature: Ambient
MDL Number: MFCD00053019
CAS Number: 2113-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR