4-Bromo-2,5-dimethoxybenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR0915-100G OR0915-25G OR0915-5G
76562-236EA 36 USD
76562-236 76560-128 76561-918
4-Bromo-2,5-dimethoxybenzaldehyde 98%
4-Bromo-2,5-dimethoxybenzaldehyde
Formula: C₉H₉BrO₃
MW: 245.08 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00156964
CAS Number: 31558-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR