3-Chloropropyldimethylammonium chloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Dimethylaminopropylchloride hydrochloride3-Dimethylaminopropyl chloride hydrochlorideN-(3-Chloropropyl)-N,N-dimethylammonium chloride3-Chloro-N,N-dimethylpropan-1-amine hydrochloride

OR0910-100G OR0910-500G
76562-070EA 16 USD
76562-070 76563-234
3-Chloropropyldimethylammonium chloride 97%
3-Chloropropyldimethylammonium chloride
Formula: C₅H₁₃Cl₂N
MW: 158.07 g/mol
Melting Pt: 138…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00012521
CAS Number: 5407-04-5
EINECS: 226-467-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR