Ethyl diethoxyacetate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Diethoxyacetic ethyl ester

OR0457-100G OR0457-25G
76563-154EA 16 USD
76563-154 76561-782
Ethyl diethoxyacetate 97%
Ethyl diethoxyacetate
Formula: C₈H₁₆O₄
MW: 176.21 g/mol
Boiling Pt: 198…200 °C
Density: 0.985 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00009231
CAS Number: 6065-82-3
EINECS: 227-999-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR