Ethyl-3,5-dichloro-4-hydroxybenzoate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Ethyl 3,5-dichloro-4-hydroxybenzoate

OR0451-1G
76563-146EA 13 USD
76563-146
Ethyl-3,5-dichloro-4-hydroxybenzoate
Ethyl 3,5-dichloro-4-hydroxybenzoate
Formula: C₉H₈Cl₂O₃
MW: 235.07 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00016420
CAS Number: 17302-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR