6-Chloropicolinic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 6-Chloropyridine-2-carboxylic acid2-Chloropyridine-6-carboxylic acid

OR0296-100G OR0296-25G
76563-152EA 69 USD
76563-152 76560-450
6-Chloropicolinic acid 98%
6-Chloropicolinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Melting Pt: 185…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00155390
CAS Number: 4684-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR