3-Amino-5-bromo-1-methyl-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

OR01988-5G OR01988-1G
76560-342EA 92 USD
76560-342 76561-682
3-Amino-5-bromo-1-methyl-1H-indazole
3-Amino-5-bromo-1-methyl-1H-indazole
Formula: C₈H₈BrN₃
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD09864804
CAS Number: 1000018-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR