Ethyl-4,6-dichloro-5-nitronicotinate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 4,6-dichloro-5-nitronicotinate

OR01969-1G
76560-920EA 123 USD
76560-920
Ethyl-4,6-dichloro-5-nitronicotinate
Ethyl-4,6-dichloro-5-nitronicotinate
Formula: C₈H₆Cl₂N₂O₄
MW: 265.05 g/mol
Melting Pt: 36…38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09842620
CAS Number: 154012-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR