Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 5-chloroimidazo[1,2-a]pyridine-2-carboxylate

OR01955-250MG OR01955-5G OR01955-1G
76563-340EA 154 USD
76563-340 76561-526 76561-230
Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 5-chloroimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₉ClN₂O₂
MW: 224.65 g/mol
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD09864797
CAS Number: 67625-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR