Methyl-3-aminosalicylate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 3-amino-2-hydroxybenzoate

OR01954-1G OR01954-100MG OR01954-250MG
76563-096EA 52 USD
76563-096 76559-654 76562-920
Methyl-3-aminosalicylate
Methyl 3-aminosalicylate
Formula: C₈H₉NO₃
MW: 167.17
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD09842614
CAS Number: 35748-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR