Imidazo[1,2-b]pyridazine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR01790-25G
76560-236EA 38 USD
76560-236
Imidazo[1,2-b]pyridazine 98%
Imidazo[1,2-b]pyridazine
Formula: C₆H₅N₃
MW: 119.12 g/mol
Melting Pt: 53…54 °C
Storage Temperature: Ambient
MDL Number: MFCD07782103
CAS Number: 766-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR