Methyl-5-phenyl-1H-pyrazole-3-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: [3-(Methoxycarbonyl)-1H-pyrazol-5-yl]benzene3-(Methoxycarbonyl)-5-phenyl-1H-pyrazole

OR01773-500MG OR01773-1G
76560-172EA 13 USD
76560-172 76563-194
Methyl-5-phenyl-1H-pyrazole-3-carboxylate
Methyl-5-phenyl-1H-pyrazole-3-carboxylate
Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00781977
CAS Number: 56426-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR