2-Amino-4,6-dimethylnicotinamide

Supplier: APOLLO SCIENTIFIC

Warning

OR017-500MG OR017-1G
76561-444EA 274 USD
76561-444 76560-974
2-Amino-4,6-dimethylnicotinamide
2-Amino-4,6-dimethylnicotinamide
Formula: C₈H₁₁N₃O
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00138572
CAS Number: 7144-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR