3-Bromoisonicotinic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Bromopyridine-4-carboxylic acid3-Bromo-4-pyridinecarboxylic acid

OR0167-5G OR0167-100G OR0167-25G
76561-412EA 110 USD
76561-412 76560-414 76563-038
3-Bromoisonicotinic acid 98%
3-Bromoisonicotinic acid
Formula: C₆H₄BrNO₂
Melting Pt: 234…238 °C
Storage Temperature: Ambient
MDL Number: MFCD00040944
CAS Number: 13959-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR