2-Bromo-5-hydroxypyrazine

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 5-Bromopyrazin-2-ol

OR01486-1G
76561-222EA 25 USD
76561-222
2-Bromo-5-hydroxypyrazine
2-Bromo-5-hydroxypyrazine
Formula: C₄H₃BrN₂O
MW: 174.9800 g/mol
Melting Pt: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD06245330
CAS Number: 374063-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR