1-Boc-piperidin-4-ylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Piperidine-4-boronic acid, pinacol ester, N-BOC protectedtert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate

45605-0050
TS45605-0050EA 433 USD
TS45605-0050
1-Boc-piperidin-4-ylboronic acid pinacol ester 97%
1-Boc-piperidin-4-ylboronic acid pinacol ester
Formula: C₁₆H₃₀BNO₄
MW: 311.2247 g/mol
Melting Pt: 56.5…57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09997792
CAS Number: 1048970-17-7

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 60 to 65 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR